Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTGAAGAGGCCGGCGCGCGTAA[A/G]GCTGTGACATTGGCCCTGGCGACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142957 MIM: 142958 MIM: 607530 MIM: 142959 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXA10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOXA10 - homeobox A10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA10-HOXA9 - HOXA10-HOXA9 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA11 - homeobox A11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA11-AS - HOXA11 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA13 - homeobox A13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |