Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAACATGCCACACGTGCTTCCCC[C/T]TCCCTGCCCCTCCTGGCCCTGTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605906 MIM: 606665 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDB3 - LIM domain binding 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080114.1 | Intron | NP_001073583.1 | ||||
NM_001080115.1 | Intron | NP_001073584.1 | ||||
NM_001080116.1 | Intron | NP_001073585.1 | ||||
NM_001171610.1 | Intron | NP_001165081.1 | ||||
NM_001171611.1 | Intron | NP_001165082.1 | ||||
NM_007078.2 | Intron | NP_009009.1 | ||||
XM_005269464.4 | Intron | XP_005269521.1 | ||||
XM_005269466.4 | Intron | XP_005269523.1 | ||||
XM_005269468.4 | Intron | XP_005269525.1 | ||||
XM_011539184.2 | Intron | XP_011537486.1 | ||||
XM_011539185.2 | Intron | XP_011537487.1 | ||||
XM_011539186.2 | Intron | XP_011537488.1 | ||||
XM_011539187.2 | Intron | XP_011537489.1 | ||||
XM_011539188.2 | Intron | XP_011537490.1 | ||||
XM_011539189.1 | Intron | XP_011537491.1 | ||||
XM_011539190.1 | Intron | XP_011537492.1 | ||||
XM_011539191.2 | Intron | XP_011537493.1 | ||||
XM_011539193.1 | Intron | XP_011537495.1 | ||||
XM_011539194.1 | Intron | XP_011537496.1 | ||||
XM_011539195.2 | Intron | XP_011537497.1 | ||||
XM_017015606.1 | Intron | XP_016871095.1 | ||||
XM_017015607.1 | Intron | XP_016871096.1 | ||||
XM_017015608.1 | Intron | XP_016871097.1 | ||||
XM_017015609.1 | Intron | XP_016871098.1 |
OPN4 - opsin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |