Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCATGGCTGCTCCTGTACGTAGT[C/T]ACGGTCTTGTGCTCTAAGGTGAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612322 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FASTKD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FASTKD2 - FAST kinase domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136193.1 | 249 | UTR 5 | NP_001129665.1 | |||
NM_001136194.1 | 249 | UTR 5 | NP_001129666.1 | |||
NM_014929.3 | 249 | UTR 5 | NP_055744.2 |
MDH1B - malate dehydrogenase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039845.2 | 249 | Intron | NP_001034934.1 | |||
NM_001282940.1 | 249 | Intron | NP_001269869.1 | |||
XM_005246315.4 | 249 | Intron | XP_005246372.1 | |||
XM_005246316.4 | 249 | Intron | XP_005246373.1 | |||
XM_011510628.2 | 249 | Intron | XP_011508930.1 | |||
XM_017003373.1 | 249 | Intron | XP_016858862.1 | |||
XM_017003374.1 | 249 | Intron | XP_016858863.1 |
Set Membership: |
HapMap |