Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACCAGGACTGGTGTCCCTCACC[C/T]TACCCCCACCCTTGCCCTCCTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606211 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD24 - ankyrin repeat domain 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133475.1 | 216 | UTR 5 | NP_597732.1 | |||
XM_011527756.2 | 216 | Intron | XP_011526058.1 | |||
XM_011527757.2 | 216 | Intron | XP_011526059.1 | |||
XM_011527758.2 | 216 | Intron | XP_011526060.1 | |||
XM_011527761.1 | 216 | Intron | XP_011526063.1 | |||
XM_011527764.2 | 216 | Intron | XP_011526066.1 | |||
XM_011527765.2 | 216 | Intron | XP_011526067.1 | |||
XM_011527768.2 | 216 | Intron | XP_011526070.1 | |||
XM_017026438.1 | 216 | Intron | XP_016881927.1 | |||
XM_017026439.1 | 216 | Intron | XP_016881928.1 | |||
XM_017026440.1 | 216 | Intron | XP_016881929.1 | |||
XM_017026441.1 | 216 | Intron | XP_016881930.1 |
SIRT6 - sirtuin 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |