Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGTCCCATGAGGGAAAAAAAC[C/T]CCGGAGCCGCAGAGAGGGGAAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610607 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPEB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPEB4 - cytoplasmic polyadenylation element binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308189.1 | 444 | UTR 5 | NP_001295118.1 | |||
NM_001308191.1 | 444 | UTR 5 | NP_001295120.1 | |||
NM_001308192.1 | 444 | Intron | NP_001295121.1 | |||
NM_001308193.1 | 444 | Intron | NP_001295122.1 | |||
NM_030627.3 | 444 | UTR 5 | NP_085130.2 | |||
XM_005265994.1 | 444 | UTR 5 | XP_005266051.1 | |||
XM_011534660.2 | 444 | UTR 5 | XP_011532962.1 | |||
XM_011534661.2 | 444 | UTR 5 | XP_011532963.1 |