Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGCGGAATAGAACAAACCAAGA[C/T]GGGTGCCCCTCCAGCTCTCTATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611758 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTUD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OTUD3 - OTU deubiquitinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015207.1 | 579 | Intron | NP_056022.1 | |||
XM_005245792.3 | 579 | Missense Mutation | ACG,ATG | T,M 1 | XP_005245849.1 | |
XM_005245793.3 | 579 | Missense Mutation | ACG,ATG | T,M 1 | XP_005245850.1 | |
XM_005245794.3 | 579 | Intron | XP_005245851.1 |