Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180465 MIM: 602671 MIM: 610971 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC84 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC84 - coiled-coil domain containing 84 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3656 - microRNA 3656 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS25 - ribosomal protein S25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001028.2 | 7 | Intron | NP_001019.1 |
SLC37A4 - solute carrier family 37 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAPPC4 - trafficking protein particle complex 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318486.1 | 7 | UTR 5 | NP_001305415.1 | |||
NM_001318488.1 | 7 | UTR 5 | NP_001305417.1 | |||
NM_001318489.1 | 7 | UTR 5 | NP_001305418.1 | |||
NM_001318490.1 | 7 | UTR 5 | NP_001305419.1 | |||
NM_001318492.1 | 7 | UTR 5 | NP_001305421.1 | |||
NM_001318494.1 | 7 | UTR 5 | NP_001305423.1 | |||
NM_016146.5 | 7 | UTR 5 | NP_057230.1 |
Set Membership: |
HapMap |