Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACGCGGGAACGCGGCCAAGCGC[A/G]CCCCATCCAGGCGGCCTATCAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603300 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00677 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00677 - long intergenic non-protein coding RNA 677 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFAIP2 - TNF alpha induced protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006291.2 | Intron | NP_006282.2 | ||||
XM_006720243.3 | Intron | XP_006720306.1 | ||||
XM_011537112.2 | Intron | XP_011535414.1 | ||||
XM_011537113.2 | Intron | XP_011535415.1 | ||||
XM_011537114.2 | Intron | XP_011535416.1 | ||||
XM_011537115.2 | Intron | XP_011535417.1 | ||||
XM_017021616.1 | Intron | XP_016877105.1 |