Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605563 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CABP1 - calcium binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033677.1 | Intron | NP_001028849.1 | ||||
NM_004276.4 | Intron | NP_004267.2 | ||||
NM_031205.3 | Intron | NP_112482.1 | ||||
XM_017020235.1 | Intron | XP_016875724.1 | ||||
XM_017020236.1 | Intron | XP_016875725.1 | ||||
XM_017020237.1 | Intron | XP_016875726.1 | ||||
XM_017020238.1 | Intron | XP_016875727.1 | ||||
XM_017020239.1 | Intron | XP_016875728.1 |