Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCCTGGACCTTCTTCTGAGGGT[A/G]GTGAGGTTTGTTTAGGGTCGCAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604196 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC27A6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC27A6 - solute carrier family 27 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017372.2 | 566 | UTR 5 | NP_001017372.1 | |||
NM_001317984.1 | 566 | UTR 5 | NP_001304913.1 | |||
NM_014031.4 | 566 | UTR 5 | NP_054750.1 |