Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGGATGGCTGCAATAGCTGGAG[A/C]TCCCATTGGTAAACCGGGTATAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604978 MIM: 615857 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDT21 - nudix hydrolase 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007006.2 | Intron | NP_008937.1 |
OGFOD1 - 2-oxoglutarate and iron dependent oxygenase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324357.1 | Intron | NP_001311286.1 | ||||
NM_001324358.1 | Intron | NP_001311287.1 | ||||
NM_001324359.1 | Intron | NP_001311288.1 | ||||
NM_001324360.1 | Intron | NP_001311289.1 | ||||
NM_001324361.1 | Intron | NP_001311290.1 | ||||
NM_001324362.1 | Intron | NP_001311291.1 | ||||
NM_001324363.1 | Intron | NP_001311292.1 | ||||
NM_018233.3 | Intron | NP_060703.3 |