Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTACTGGCAGAGAGAGCTTTGGTA[C/G]GGCTATTGCTATTTAAAAACCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176312 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC51145 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC51145 - uncharacterized LOC51145 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PBX3 - PBX homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134778.1 | Intron | NP_001128250.1 | ||||
NM_006195.5 | Intron | NP_006186.1 | ||||
XM_006717130.1 | Intron | XP_006717193.1 | ||||
XM_006717131.1 | Intron | XP_006717194.1 | ||||
XM_006717132.1 | Intron | XP_006717195.1 | ||||
XM_006717133.1 | Intron | XP_006717196.1 | ||||
XM_011518755.1 | Intron | XP_011517057.1 | ||||
XM_017014786.1 | Intron | XP_016870275.1 |