Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAAATTTTCGATAATCTATTCT[C/T]GGGATCGTCTTTTGTTTCTTCAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610450 MIM: 608789 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYPD1 - LY6/PLAUR domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCKAP5 - NCK associated protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207363.2 | 6124 | UTR 3 | NP_997246.2 | |||
NM_207481.3 | 6124 | UTR 3 | NP_997364.3 | |||
XM_005263659.3 | 6124 | Missense Mutation | AAG,GAG | K,E 1873 | XP_005263716.1 | |
XM_005263660.3 | 6124 | Missense Mutation | AAG,GAG | K,E 1873 | XP_005263717.1 | |
XM_011511097.2 | 6124 | Missense Mutation | AAG,GAG | K,E 1873 | XP_011509399.1 | |
XM_011511098.2 | 6124 | Missense Mutation | AAG,GAG | K,E 1873 | XP_011509400.1 | |
XM_011511099.2 | 6124 | Missense Mutation | AAG,GAG | K,E 1873 | XP_011509401.1 | |
XM_011511100.2 | 6124 | Missense Mutation | AAG,GAG | K,E 1873 | XP_011509402.1 | |
XM_011511101.1 | 6124 | Missense Mutation | AAG,GAG | K,E 1848 | XP_011509403.1 | |
XM_011511102.2 | 6124 | Missense Mutation | AAG,GAG | K,E 1848 | XP_011509404.1 | |
XM_011511103.2 | 6124 | Missense Mutation | AAG,GAG | K,E 1805 | XP_011509405.1 | |
XM_011511104.1 | 6124 | Missense Mutation | AAG,GAG | K,E 1805 | XP_011509406.1 | |
XM_011511105.2 | 6124 | Missense Mutation | AAG,GAG | K,E 1805 | XP_011509407.1 | |
XM_017003974.1 | 6124 | Missense Mutation | AAG,GAG | K,E 1873 | XP_016859463.1 | |
XM_017003975.1 | 6124 | Missense Mutation | AAG,GAG | K,E 1873 | XP_016859464.1 | |
XM_017003976.1 | 6124 | Missense Mutation | AAG,GAG | K,E 1848 | XP_016859465.1 | |
XM_017003977.1 | 6124 | Missense Mutation | AAG,GAG | K,E 1848 | XP_016859466.1 | |
XM_017003978.1 | 6124 | Missense Mutation | AAG,GAG | K,E 1712 | XP_016859467.1 | |
XM_017003979.1 | 6124 | Missense Mutation | AAG,GAG | K,E 554 | XP_016859468.1 | |
XM_017003980.1 | 6124 | UTR 3 | XP_016859469.1 |