Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGTGGCAAGCCAGACCTGTGGCT[C/G]AGGGCCAGTATCCCTCCACCAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609799 MIM: 602464 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM222B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM222B - family with sequence similarity 222 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEK8 - NIMA related kinase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRAF4 - TNF receptor associated factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004295.3 | Intron | NP_004286.2 | ||||
XM_011525504.2 | Intron | XP_011523806.1 | ||||
XM_011525505.2 | Intron | XP_011523807.1 | ||||
XM_011525506.2 | Intron | XP_011523808.1 | ||||
XM_011525507.2 | Intron | XP_011523809.1 | ||||
XM_011525508.2 | Intron | XP_011523810.1 |