Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 138320 MIM: 165390 | |||||||||||||||||||||||
Literature Links: |
GPX1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GPX1 - glutathione peroxidase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOA - ras homolog family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313941.1 | 1097 | UTR 3 | NP_001300870.1 | |||
NM_001313943.1 | 1097 | UTR 3 | NP_001300872.1 | |||
NM_001313944.1 | 1097 | UTR 3 | NP_001300873.1 | |||
NM_001313945.1 | 1097 | UTR 3 | NP_001300874.1 | |||
NM_001313946.1 | 1097 | UTR 3 | NP_001300875.1 | |||
NM_001313947.1 | 1097 | UTR 3 | NP_001300876.1 | |||
NM_001664.3 | 1097 | UTR 3 | NP_001655.1 |