Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGCTTGTGAGTGGGGCTGCCG[A/G]GAGGGCAGGCGTGGGGCGAGGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC173 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC173 - coiled-coil domain containing 173 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085447.1 | 86 | Intron | NP_001078916.1 | |||
XM_011510589.1 | 86 | Intron | XP_011508891.1 | |||
XM_011510590.1 | 86 | Intron | XP_011508892.1 |
PHOSPHO2 - phosphatase, orphan 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008489.3 | 86 | UTR 5 | NP_001008489.1 | |||
NM_001199285.1 | 86 | Intron | NP_001186214.1 | |||
NM_001199286.1 | 86 | UTR 5 | NP_001186215.1 | |||
NM_001199287.1 | 86 | UTR 5 | NP_001186216.1 | |||
NM_001199288.1 | 86 | UTR 5 | NP_001186217.1 |
PHOSPHO2-KLHL23 - PHOSPHO2-KLHL23 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199290.1 | 86 | UTR 5 | NP_001186219.1 |