Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCAGGCTGCGCTGCCCGCAGTA[C/T]TGGACCCGAGCGCGACGGTGCGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616189 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMTR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CMTR1 - cap methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015050.2 | 97 | UTR 5 | NP_055865.1 | |||
XM_005248955.3 | 97 | UTR 5 | XP_005249012.1 |
LOC107986531 - oleosin-B6-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017011595.1 | 97 | Missense Mutation | ATA,GTA | I,V 120 | XP_016867084.1 |