Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCACTGCCAGCCTGTGTATCCCC[G/T]TTGGCACCCCGCCCAAACGGGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165220 MIM: 601233 MIM: 612031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLI1 - GLI family zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160045.1 | Intron | NP_001153517.1 | ||||
NM_001167609.1 | Intron | NP_001161081.1 | ||||
NM_005269.2 | Intron | NP_005260.1 | ||||
XM_011538189.2 | Intron | XP_011536491.1 | ||||
XM_011538190.1 | Intron | XP_011536492.1 |
INHBC - inhibin beta C subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
INHBE - inhibin beta E subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |