Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604693 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AKAP7 - A-kinase anchoring protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004842.3 | 109 | Intron | NP_004833.1 | |||
NM_016377.3 | 109 | Missense Mutation | AAA,GAA | K,E 26 | NP_057461.2 | |
NM_138633.2 | 109 | Intron | NP_619539.1 | |||
XM_005267228.1 | 109 | Missense Mutation | AAA,GAA | K,E 26 | XP_005267285.1 | |
XM_005267229.4 | 109 | Missense Mutation | AAA,GAA | K,E 4 | XP_005267286.1 | |
XM_006715614.2 | 109 | Missense Mutation | AAA,GAA | K,E 26 | XP_006715677.1 | |
XM_011536262.1 | 109 | Missense Mutation | AAA,GAA | K,E 26 | XP_011534564.1 | |
XM_017011506.1 | 109 | Missense Mutation | AAA,GAA | K,E 26 | XP_016866995.1 | |
XM_017011507.1 | 109 | Missense Mutation | AAA,GAA | K,E 26 | XP_016866996.1 | |
XM_017011508.1 | 109 | Missense Mutation | AAA,GAA | K,E 36 | XP_016866997.1 | |
XM_017011509.1 | 109 | Missense Mutation | AAA,GAA | K,E 30 | XP_016866998.1 | |
XM_017011510.1 | 109 | Intron | XP_016866999.1 | |||
XM_017011511.1 | 109 | Intron | XP_016867000.1 |
Set Membership: |
HapMap |