Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K7CL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K7CL - MAP3K7 C-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286617.1 | Intron | NP_001273546.1 | ||||
NM_001286618.1 | Intron | NP_001273547.1 | ||||
NM_001286619.1 | Intron | NP_001273548.1 | ||||
NM_001286620.1 | Intron | NP_001273549.1 | ||||
NM_001286622.1 | Intron | NP_001273551.1 | ||||
NM_001286623.1 | Intron | NP_001273552.1 | ||||
NM_001286624.1 | Intron | NP_001273553.1 | ||||
NM_001286634.1 | Intron | NP_001273563.1 | ||||
NM_020152.3 | Intron | NP_064537.1 |