Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGCGAGGGGGCGGCGAATGGCG[C/T]GCGGTCGGAGAGTACGAGCGCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611230 MIM: 604272 MIM: 131222 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCAPH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCAPH2 - non-SMC condensin II complex subunit H2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ODF3B - outer dense fiber of sperm tails 3B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCO2 - SCO2 cytochrome c oxidase assembly protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169109.1 | 1620 | Intron | NP_001162580.1 | |||
NM_001169110.1 | 1620 | Intron | NP_001162581.1 | |||
NM_001169111.1 | 1620 | Intron | NP_001162582.1 | |||
NM_005138.2 | 1620 | Intron | NP_005129.2 |
TYMP - thymidine phosphorylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113755.2 | 1620 | Missense Mutation | ACG,GCG | T,A 465 | NP_001107227.1 | |
NM_001113756.2 | 1620 | Missense Mutation | ACG,GCG | T,A 465 | NP_001107228.1 | |
NM_001257988.1 | 1620 | Missense Mutation | ACG,GCG | T,A 465 | NP_001244917.1 | |
NM_001257989.1 | 1620 | Missense Mutation | ACG,GCG | T,A 470 | NP_001244918.1 | |
NM_001953.4 | 1620 | Missense Mutation | ACG,GCG | T,A 465 | NP_001944.1 |