Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTGTGCCAATCCATTGTCCACC[C/T]CTTTGAGGATGACATGGTGGAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602709 MIM: 607608 | ||||||||||||||||||||
Literature Links: |
APBB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APBB1 - amyloid beta precursor protein binding family B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMPD1 - sphingomyelin phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000543.4 | 601 | Missense Mutation | CCC,CTC | P,L 139 | NP_000534.3 | |
NM_001007593.2 | 601 | Missense Mutation | CCC,CTC | P,L 138 | NP_001007594.2 | |
NM_001318087.1 | 601 | Missense Mutation | CCC,CTC | P,L 139 | NP_001305016.1 | |
NM_001318088.1 | 601 | UTR 5 | NP_001305017.1 | |||
XM_011520303.1 | 601 | Missense Mutation | CCC,CTC | P,L 139 | XP_011518605.1 | |
XM_011520304.1 | 601 | Missense Mutation | CCC,CTC | P,L 139 | XP_011518606.1 |