Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGGCACAGTCGTAGCAGGGGCTC[C/T]AGGAGGTGAACCAGGTGACGCGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605257 | ||||||||||||||||||||
Literature Links: |
AICDA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AICDA - activation-induced cytidine deaminase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020661.2 | 449 | Nonsense Mutation | NP_065712.1 | |||
XM_011520772.2 | 449 | Nonsense Mutation | TAG,TGG | *,W 84 | XP_011519074.1 | |
XM_011520773.2 | 449 | Nonsense Mutation | TAG,TGG | *,W 84 | XP_011519075.1 |