Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGAGGTCATGAAACCGCCGTAGC[C/G]CTTGTCCTTGGGCGGGCTGCCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616797 MIM: 176830 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFR3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFR3B - EFR3 homolog B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POMC - proopiomelanocortin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000939.3 | 1110 | Missense Mutation | CGC,GGC | R,G 236 | NP_000930.1 | |
NM_001035256.2 | 1110 | Missense Mutation | CGC,GGC | R,G 236 | NP_001030333.1 | |
NM_001319204.1 | 1110 | Missense Mutation | CGC,GGC | R,G 236 | NP_001306133.1 | |
NM_001319205.1 | 1110 | Missense Mutation | CGC,GGC | R,G 236 | NP_001306134.1 |