Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGGCGGGATCGACTGGGCTCTTA[C/G]AGCGGCCCCACCTCGGTCTCCCGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606157 | ||||||||||||||||||||
Literature Links: |
PANK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PANK2 - pantothenate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324191.1 | 737 | UTR 5 | NP_001311120.1 | |||
NM_001324192.1 | 737 | Nonsense Mutation | TAC,TAG | Y,* 190 | NP_001311121.1 | |
NM_001324193.1 | 737 | Intron | NP_001311122.1 | |||
NM_024960.5 | 737 | Intron | NP_079236.3 | |||
NM_153638.3 | 737 | Nonsense Mutation | TAC,TAG | Y,* 190 | NP_705902.2 | |
NM_153640.3 | 737 | Intron | NP_705904.1 | |||
XM_005260835.3 | 737 | Intron | XP_005260892.1 | |||
XM_005260836.4 | 737 | Intron | XP_005260893.3 | |||
XM_011529364.2 | 737 | Nonsense Mutation | TAC,TAG | Y,* 190 | XP_011527666.1 | |
XM_011529365.2 | 737 | Nonsense Mutation | TAC,TAG | Y,* 190 | XP_011527667.1 | |
XM_017028077.1 | 737 | Intron | XP_016883566.1 | |||
XM_017028078.1 | 737 | Intron | XP_016883567.1 | |||
XM_017028079.1 | 737 | Intron | XP_016883568.1 |