Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTTATCCACTTACCAAAAGCCC[G/A]AATGTCAGATTTGCTGCTGAACTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300300 MIM: 300356 | ||||||||||||||||||||
Literature Links: |
BTK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTK - Bruton tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000061.2 | 1934 | Missense Mutation | CGG,TGG | R,W 581 | NP_000052.1 | |
NM_001287344.1 | 1934 | Missense Mutation | CGG,TGG | R,W 615 | NP_001274273.1 | |
NM_001287345.1 | 1934 | Missense Mutation | CGG,TGG | R,W 405 | NP_001274274.1 |
TIMM8A - translocase of inner mitochondrial membrane 8 homolog A (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |