Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCCCGCCAAGGTTCTGGAATCC[C/T]GGAGGTGAAGACCATGTTGGCGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602023 | ||||||||||||||||||||
Literature Links: |
CLCNKB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLCNKB - chloride voltage-gated channel Kb | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000085.4 | 523 | Missense Mutation | CCG,CTG | P,L 124 | NP_000076.2 | |
NM_001165945.2 | 523 | Intron | NP_001159417.2 |
FAM131C - family with sequence similarity 131 member C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |