Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCCTGACCGTAATCCCCAGGGT[C/T]GAGCCGTGCTGCACGTGGCTCTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 172400 | ||||||||||||||||||||
Literature Links: |
GPI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPI - glucose-6-phosphate isomerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184722.1 | 343 | Intron | NP_001171651.1 | |||
NM_001289789.1 | 343 | Intron | NP_001276718.1 | |||
XM_005258764.2 | 343 | Nonsense Mutation | CGA,TGA | R,* 96 | XP_005258821.1 | |
XM_006723148.2 | 343 | Nonsense Mutation | CGA,TGA | R,* 96 | XP_006723211.1 | |
XM_011526754.2 | 343 | Nonsense Mutation | CGA,TGA | R,* 135 | XP_011525056.1 |