Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTTCAAACAGATCACATCCACA[T/C]ACGGAGCCTAGGACCAGAGCAGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611499 MIM: 608838 | ||||||||||||||||||||
Literature Links: |
GUSB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GUSB - glucuronidase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000181.3 | 1359 | Missense Mutation | TAT,TGT | Y,C 495 | NP_000172.2 | |
NM_001284290.1 | 1359 | Missense Mutation | TAT,TGT | Y,C 349 | NP_001271219.1 | |
NM_001293104.1 | 1359 | Missense Mutation | TAT,TGT | Y,C 305 | NP_001280033.1 | |
NM_001293105.1 | 1359 | Missense Mutation | TAT,TGT | Y,C 276 | NP_001280034.1 | |
XM_005250297.3 | 1359 | Missense Mutation | TAT,TGT | Y,C 444 | XP_005250354.1 | |
XM_011516113.1 | 1359 | Missense Mutation | TAT,TGT | Y,C 328 | XP_011514415.1 | |
XM_011516114.1 | 1359 | Missense Mutation | TAT,TGT | Y,C 271 | XP_011514416.1 | |
XM_017012091.1 | 1359 | Missense Mutation | TAT,TGT | Y,C 277 | XP_016867580.1 | |
XM_017012092.1 | 1359 | Missense Mutation | TAT,TGT | Y,C 254 | XP_016867581.1 | |
XM_017012093.1 | 1359 | Missense Mutation | TAT,TGT | Y,C 220 | XP_016867582.1 |
VKORC1L1 - vitamin K epoxide reductase complex subunit 1 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |