Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTACCTGACAATGACGCCCGGC[C/T]GGAGGTCGAAGTTCTTATGCACCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAT1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAT1A - methionine adenosyltransferase 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000429.2 | 1322 | Missense Mutation | NP_000420.1 | |||
XM_005269842.4 | 1322 | Missense Mutation | XP_005269899.1 | |||
XM_005269843.4 | 1322 | Missense Mutation | XP_005269900.1 |