Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCAATCATTAAGAAGACAAAGG[A/G]TTTGTTGAACTTGACCTCGGGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107400 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPINA1 - serpin family A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000295.4 | 1710 | Missense Mutation | NP_000286.3 | |||
NM_001002235.2 | 1710 | Missense Mutation | NP_001002235.1 | |||
NM_001002236.2 | 1710 | Missense Mutation | NP_001002236.1 | |||
NM_001127700.1 | 1710 | Missense Mutation | NP_001121172.1 | |||
NM_001127701.1 | 1710 | Missense Mutation | NP_001121173.1 | |||
NM_001127702.1 | 1710 | Missense Mutation | NP_001121174.1 | |||
NM_001127703.1 | 1710 | Missense Mutation | NP_001121175.1 | |||
NM_001127704.1 | 1710 | Missense Mutation | NP_001121176.1 | |||
NM_001127705.1 | 1710 | Missense Mutation | NP_001121177.1 | |||
NM_001127706.1 | 1710 | Missense Mutation | NP_001121178.1 | |||
NM_001127707.1 | 1710 | Missense Mutation | NP_001121179.1 | |||
XM_017021370.1 | 1710 | Missense Mutation | XP_016876859.1 |