Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCAATGTGCAGCACAGCCTTAT[A/G]GGCAGCCTGTGTGGGGAGAACAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 314200 | ||||||||||||||||||||
Literature Links: |
SERPINA7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERPINA7 - serpin family A member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000354.5 | 1157 | Missense Mutation | CAT,TAT | H,Y 351 | NP_000345.2 | |
XM_005262180.3 | 1157 | Missense Mutation | CCA,CTA | P,L 371 | XP_005262237.1 | |
XM_006724683.2 | 1157 | Missense Mutation | CAT,TAT | H,Y 361 | XP_006724746.1 |