Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610737 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KSR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KSR2 - kinase suppressor of ras 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173598.4 | 12896 | UTR 3 | NP_775869.3 | |||
XM_011538224.2 | 12896 | UTR 3 | XP_011536526.1 | |||
XM_011538225.2 | 12896 | UTR 3 | XP_011536527.1 | |||
XM_011538226.2 | 12896 | Intron | XP_011536528.1 | |||
XM_011538228.2 | 12896 | UTR 3 | XP_011536530.1 | |||
XM_011538229.2 | 12896 | Intron | XP_011536531.1 | |||
XM_017019208.1 | 12896 | Intron | XP_016874697.1 | |||
XM_017019209.1 | 12896 | Intron | XP_016874698.1 | |||
XM_017019210.1 | 12896 | UTR 3 | XP_016874699.1 |
Set Membership: |
HapMap |