Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600003 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CACNB2 - calcium voltage-gated channel auxiliary subunit beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NSUN6 - NOP2/Sun RNA methyltransferase family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182543.3 | Intron | NP_872349.1 | ||||
XM_011519383.1 | Intron | XP_011517685.1 | ||||
XM_011519384.1 | Intron | XP_011517686.1 | ||||
XM_011519385.1 | Intron | XP_011517687.1 | ||||
XM_011519386.2 | Intron | XP_011517688.1 | ||||
XM_011519387.2 | Intron | XP_011517689.1 | ||||
XM_011519389.2 | Intron | XP_011517691.1 | ||||
XM_017015906.1 | Intron | XP_016871395.1 | ||||
XM_017015907.1 | Intron | XP_016871396.1 | ||||
XM_017015908.1 | Intron | XP_016871397.1 | ||||
XM_017015909.1 | Intron | XP_016871398.1 | ||||
XM_017015910.1 | Intron | XP_016871399.1 | ||||
XM_017015911.1 | Intron | XP_016871400.1 | ||||
XM_017015912.1 | Intron | XP_016871401.1 | ||||
XM_017015913.1 | Intron | XP_016871402.1 | ||||
XM_017015914.1 | Intron | XP_016871403.1 |