Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 167416 MIM: 607571 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAX9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAX9 - paired box 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A21 - solute carrier family 25 member 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171170.1 | 2034 | Intron | NP_001164641.1 | |||
NM_030631.3 | 2034 | UTR 3 | NP_085134.1 | |||
XM_011537287.2 | 2034 | Intron | XP_011535589.1 | |||
XM_011537288.2 | 2034 | Intron | XP_011535590.1 | |||
XM_011537289.2 | 2034 | Intron | XP_011535591.1 |
Set Membership: |
HapMap |