Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606620 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf204 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf204 - chromosome 1 open reading frame 204 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134233.1 | 2112 | UTR 3 | NP_001127705.1 |
SLAMF8 - SLAM family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020125.2 | 2112 | Intron | NP_064510.1 | |||
XM_005245345.2 | 2112 | Intron | XP_005245402.1 | |||
XM_011509775.1 | 2112 | Intron | XP_011508077.1 | |||
XM_011509776.2 | 2112 | Intron | XP_011508078.1 | |||
XM_011509777.2 | 2112 | UTR 3 | XP_011508079.1 | |||
XM_011509778.2 | 2112 | Intron | XP_011508080.1 | |||
XM_011509779.2 | 2112 | Intron | XP_011508081.1 | |||
XM_011509780.2 | 2112 | Intron | XP_011508082.2 | |||
XM_017001844.1 | 2112 | UTR 3 | XP_016857333.1 | |||
XM_017001845.1 | 2112 | Intron | XP_016857334.1 | |||
XM_017001846.1 | 2112 | Intron | XP_016857335.1 |
Set Membership: |
HapMap |