Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612188 | ||||||||||||||||||||
Literature Links: |
PLA2G4F PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLA2G4F - phospholipase A2 group IVF | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS39 - VPS39, HOPS complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301138.1 | 2594 | Missense Mutation | GTC,TTC | V,F 817 | NP_001288067.1 | |
NM_015289.3 | 2594 | Missense Mutation | GTC,TTC | V,F 806 | NP_056104.2 | |
XM_011521403.2 | 2594 | Missense Mutation | GTC,TTC | V,F 817 | XP_011519705.1 | |
XM_011521404.2 | 2594 | Missense Mutation | GTC,TTC | V,F 806 | XP_011519706.1 | |
XM_017022036.1 | 2594 | Missense Mutation | GTC,TTC | V,F 817 | XP_016877525.1 | |
XM_017022037.1 | 2594 | Missense Mutation | GTC,TTC | V,F 806 | XP_016877526.1 | |
XM_017022038.1 | 2594 | Missense Mutation | GTC,TTC | V,F 682 | XP_016877527.1 | |
XM_017022039.1 | 2594 | Missense Mutation | GTC,TTC | V,F 449 | XP_016877528.1 |