Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602620 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGMN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LGMN - legumain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008530.2 | Intron | NP_001008530.1 | ||||
NM_005606.6 | Intron | NP_005597.3 | ||||
XM_005267862.3 | Intron | XP_005267919.1 | ||||
XM_005267863.3 | Intron | XP_005267920.1 | ||||
XM_011536970.1 | Intron | XP_011535272.1 | ||||
XM_017021463.1 | Intron | XP_016876952.1 | ||||
XM_017021464.1 | Intron | XP_016876953.1 | ||||
XM_017021465.1 | Intron | XP_016876954.1 | ||||
XM_017021466.1 | Intron | XP_016876955.1 | ||||
XM_017021467.1 | Intron | XP_016876956.1 |
Set Membership: |
HapMap |