Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATGAACCATGTCTTTAGTTGG[A/G]GAACCATTTGTCTGAAATGAATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605826 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM234B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM234B - family with sequence similarity 234 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020853.1 | Intron | NP_065904.1 | ||||
XM_017019706.1 | Intron | XP_016875195.1 |
HEBP1 - heme binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015987.4 | Intron | NP_057071.2 |
LOC100506314 - uncharacterized LOC100506314 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |