Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605140 | ||||||||||||||||||||
Literature Links: |
CCT7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCT7 - chaperonin containing TCP1 subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009570.2 | Intron | NP_001009570.1 | ||||
NM_001166284.1 | Intron | NP_001159756.1 | ||||
NM_001166285.1 | Intron | NP_001159757.1 | ||||
NM_006429.3 | Intron | NP_006420.1 |
PRADC1 - protease associated domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |