Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615497 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIEF1 - mitochondrial elongation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304564.1 | Intron | NP_001291493.1 | ||||
NM_019008.5 | Intron | NP_061881.2 | ||||
XM_011530236.1 | Intron | XP_011528538.1 | ||||
XM_011530237.1 | Intron | XP_011528539.1 | ||||
XM_011530238.1 | Intron | XP_011528540.1 | ||||
XM_017028838.1 | Intron | XP_016884327.1 |