Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606741 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF181 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF181 - zinc finger protein 181 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029997.3 | 3265 | Intron | NP_001025168.2 | |||
NM_001145665.1 | 3265 | Intron | NP_001139137.1 | |||
XM_005258849.2 | 3265 | Intron | XP_005258906.1 | |||
XM_005258850.2 | 3265 | Intron | XP_005258907.1 | |||
XM_005258851.3 | 3265 | UTR 3 | XP_005258908.1 | |||
XM_006723183.3 | 3265 | Intron | XP_006723246.1 | |||
XM_006723184.3 | 3265 | UTR 3 | XP_006723247.1 | |||
XM_017026739.1 | 3265 | Intron | XP_016882228.1 | |||
XM_017026740.1 | 3265 | UTR 3 | XP_016882229.1 |