Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604048 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC22A14 - solute carrier family 22 member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320033.1 | Intron | NP_001306962.1 | ||||
NM_004803.4 | Intron | NP_004794.2 | ||||
XM_005265585.4 | Intron | XP_005265642.1 | ||||
XM_006713416.3 | Intron | XP_006713479.2 | ||||
XM_006713417.3 | Intron | XP_006713480.2 | ||||
XM_006713418.3 | Intron | XP_006713481.2 | ||||
XM_011534245.2 | Intron | XP_011532547.1 | ||||
XM_011534247.2 | Intron | XP_011532549.1 |
Set Membership: |
HapMap |