Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189971 MIM: 612478 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTL10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACTL10 - actin like 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024675.1 | 1983 | UTR 3 | NP_001019846.1 |
C20orf144 - chromosome 20 open reading frame 144 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
E2F1 - E2F transcription factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NECAB3 - N-terminal EF-hand calcium binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031231.3 | 1983 | Intron | NP_112508.3 | |||
NM_031232.3 | 1983 | Intron | NP_112509.3 | |||
XM_005260510.1 | 1983 | Intron | XP_005260567.1 | |||
XM_011528991.1 | 1983 | Intron | XP_011527293.1 | |||
XM_011528992.2 | 1983 | Intron | XP_011527294.1 | |||
XM_017028015.1 | 1983 | Intron | XP_016883504.1 | |||
XM_017028016.1 | 1983 | Intron | XP_016883505.1 |
Set Membership: |
HapMap |