Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604889 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NBEA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NBEA - neurobeachin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204197.1 | Intron | NP_001191126.1 | ||||
NM_015678.4 | Intron | NP_056493.3 | ||||
XM_005266346.4 | Intron | XP_005266403.1 | ||||
XM_005266347.4 | Intron | XP_005266404.2 | ||||
XM_005266348.4 | Intron | XP_005266405.2 | ||||
XM_006719803.3 | Intron | XP_006719866.1 | ||||
XM_006719805.3 | Intron | XP_006719868.1 | ||||
XM_006719806.3 | Intron | XP_006719869.1 | ||||
XM_011535046.2 | Intron | XP_011533348.1 | ||||
XM_011535047.2 | Intron | XP_011533349.1 | ||||
XM_017020544.1 | Intron | XP_016876033.1 | ||||
XM_017020545.1 | Intron | XP_016876034.1 | ||||
XM_017020546.1 | Intron | XP_016876035.1 |