Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605085 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TREM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TREM1 - triggering receptor expressed on myeloid cells 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242589.2 | 4662 | Intron | NP_001229518.1 | |||
NM_001242590.2 | 4662 | Intron | NP_001229519.1 | |||
NM_018643.4 | 4662 | Intron | NP_061113.1 | |||
XM_006715117.3 | 4662 | Intron | XP_006715180.1 | |||
XM_011514696.1 | 4662 | Intron | XP_011512998.1 | |||
XM_011514697.1 | 4662 | Intron | XP_011512999.1 | |||
XM_017010956.1 | 4662 | UTR 3 | XP_016866445.1 | |||
XM_017010957.1 | 4662 | UTR 3 | XP_016866446.1 |