Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600909 MIM: 612412 | ||||||||||||||||||||
Literature Links: |
LSS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LSS - lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001438.2 | 2577 | UTR 3 | NP_001001438.1 | |||
NM_001145436.1 | 2577 | Intron | NP_001138908.1 | |||
NM_001145437.1 | 2577 | Intron | NP_001138909.1 | |||
NM_002340.5 | 2577 | Intron | NP_002331.3 |
SPATC1L - spermatogenesis and centriole associated 1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |