Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCCTTCTGCAAAACCCCACTGC[A/G]CCTATTAGTCTTTATGCATCTATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607139 MIM: 609217 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FANCA - Fanconi anemia complementation group A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107984817 - uncharacterized LOC107984817 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPIRE2 - spire type actin nucleation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032451.1 | Intron | NP_115827.1 | ||||
XM_011523379.1 | Intron | XP_011521681.1 | ||||
XM_011523380.2 | Intron | XP_011521682.1 | ||||
XM_011523381.1 | Intron | XP_011521683.1 | ||||
XM_011523384.1 | Intron | XP_011521686.1 | ||||
XM_011523385.1 | Intron | XP_011521687.1 | ||||
XM_017023779.1 | Intron | XP_016879268.1 | ||||
XM_017023780.1 | Intron | XP_016879269.1 | ||||
XM_017023781.1 | Intron | XP_016879270.1 |
Set Membership: |
HapMap |