Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A31 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC22A31 - solute carrier family 22 member 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF778 - zinc finger protein 778 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201407.1 | Intron | NP_001188336.1 | ||||
NM_182531.3 | Intron | NP_872337.2 | ||||
XM_005256288.2 | Intron | XP_005256345.1 | ||||
XM_011522940.2 | Intron | XP_011521242.1 | ||||
XM_017023015.1 | Intron | XP_016878504.1 | ||||
XM_017023016.1 | Intron | XP_016878505.1 | ||||
XM_017023017.1 | Intron | XP_016878506.1 |
Set Membership: |
HapMap |