Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615855 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OVCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OVCH1 - ovochymase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMTC1 - transmembrane and tetratricopeptide repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193451.1 | 7124 | UTR 3 | NP_001180380.1 | |||
NM_175861.3 | 7124 | UTR 3 | NP_787057.2 | |||
XM_005253499.4 | 7124 | UTR 3 | XP_005253556.1 | |||
XM_017020003.1 | 7124 | UTR 3 | XP_016875492.1 | |||
XM_017020004.1 | 7124 | UTR 3 | XP_016875493.1 | |||
XM_017020005.1 | 7124 | UTR 3 | XP_016875494.1 | |||
XM_017020006.1 | 7124 | Intron | XP_016875495.1 | |||
XM_017020007.1 | 7124 | UTR 3 | XP_016875496.1 |